ENAH5802EB58 Enamine

(1E)-1-ethoxy-4,4-difluoropent-1-en-3-one

  • MDL number MFCD18810806
  • Cas Number : 461451-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7438504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438504
Availability details for
Quantity
  • Availability
  • Add to Cart