ENAH5802EB5B Enamine

1,3-dimethyl-1H,4H,5H,6H,7H-pyrazolo[3,4-b]pyridin-4-one

  • MDL number MFCD30619760
  • Cas Number : 2169008-15-3
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dimethyl-1,5,6,7-tetrahydro-4H-pyrazolo[3,4-b]pyridin-4-one
Inchi Code 1S/C8H11N3O/c1-5-7-6(12)3-4-9-8(7)11(2)10-5/h9H,3-4H2,1-2H3
Inchi Key DFIMEJHWGKCMJJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-305947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-305947
Availability details for
Quantity
  • Availability
  • Add to Cart