ENAH5802EB5E Enamine

3-azaspiro[5.5]undecan-9-one hydrochloride

  • MDL number MFCD27920789
  • Cas Number : 1225437-09-1
  • Molecular Weight : 203.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7353335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353335
Availability details for
Quantity
  • Availability
  • Add to Cart