ENAH5802EB5F Enamine

1,5-dimethyl-9-azabicyclo[3.3.1]nonan-3-one hydrochloride

  • MDL number MFCD32065966
  • Cas Number : 2361644-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431324
Availability details for
Quantity
  • Availability
  • Add to Cart