ENAH5802EB60 Enamine

4-(pyrrolidin-1-yl)cyclohexan-1-one hydrochloride

  • MDL number MFCD32065923
  • Cas Number : 2361644-21-3
  • Molecular Weight : 203.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(pyrrolidin-1-yl)cyclohexan-1-one hydrochloride
Inchi Code 1S/C10H17NO.ClH/c12-10-5-3-9(4-6-10)11-7-1-2-8-11;/h9H,1-8H2;1H
Inchi Key ZAAUPXPDWWAAMG-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430961
Availability details for
Quantity
  • Availability
  • Add to Cart