ENAH5802EB65 Enamine

1-(4,4-dimethylcycloheptyl)ethan-1-one

  • MDL number MFCD21775967
  • Cas Number : 1507473-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3000628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3000628
Availability details for
Quantity
  • Availability
  • Add to Cart