ENAH5802EB66 Enamine

5-silaspiro[4.5]decan-8-one

  • MDL number MFCD24712988
  • Cas Number : 1330166-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-6746677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746677
Availability details for
Quantity
  • Availability
  • Add to Cart