ENAH5802EB69 Enamine

1-(5-nitrothiophen-3-yl)ethan-1-one

  • MDL number MFCD00084862
  • Cas Number : 58157-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-62
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-6739751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739751
Availability details for
Quantity
  • Availability
  • Add to Cart