ENAH5802EB6B Enamine

ethyl (3E)-4-ethoxy-2-oxobut-3-enoate

  • Cas Number : 65260-58-4
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (3E)-4-ethoxy-2-oxo-3-butenoate
Inchi Code 1S/C8H12O4/c1-3-11-6-5-7(9)8(10)12-4-2/h5-6H,3-4H2,1-2H3/b6-5+
Inchi Key XPBYGYFJOZXYOE-AATRIKPKSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1198115

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1198115
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart