ENAH5802EB72 Enamine

2-phenoxycyclopentan-1-one

  • MDL number MFCD24388003
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-phenoxycyclopentan-1-one
Inchi Code 1S/C11H12O2/c12-10-7-4-8-11(10)13-9-5-2-1-3-6-9/h1-3,5-6,11H,4,7-8H2
Inchi Key BLJQDYYIDRRVOZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-171226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171226
Availability details for
Quantity
  • Availability
  • Add to Cart