ENAH5802EB75 Enamine

1-(1-benzothiophen-5-yl)ethan-1-one

  • MDL number MFCD16619666
  • Cas Number : 1128-88-7
  • Molecular Weight : 176.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1-benzothien-5-yl)ethanone
Inchi Code 1S/C10H8OS/c1-7(11)8-2-3-10-9(6-8)4-5-12-10/h2-6H,1H3
Inchi Key LYPSYIWSZQVHAB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2057499

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart