ENAH5802EB77 Enamine

4-(phenylsulfanyl)but-3-en-2-one

  • MDL number MFCD30232034
  • Cas Number : 92897-11-5
  • Molecular Weight : 178.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 30-31
Iupac Name (E)-4-(phenylthio)but-3-en-2-one
Inchi Code 1S/C10H10OS/c1-9(11)7-8-12-10-5-3-2-4-6-10/h2-8H,1H3/b8-7+
Inchi Key IUDATFOKAOFQMH-BQYQJAHWSA-N

Description

Melting Point 30-31
Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430096

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7430096
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart