ENAH5802EB7B Enamine

1,5,7-trimethylbicyclo[3.3.1]nonan-3-one

  • MDL number MFCD32069085
  • Cas Number : 1934982-87-2
  • Molecular Weight : 180.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,5,7-trimethylbicyclo[3.3.1]nonan-3-one
Inchi Code 1S/C12H20O/c1-9-4-11(2)6-10(13)7-12(3,5-9)8-11/h9H,4-8H2,1-3H3
Inchi Key ZFZWQPRCPYAFHS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55003

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-55003
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart