ENAH5802EB7E Enamine

methyl 6-oxospiro[2.5]octane-1-carboxylate

  • MDL number MFCD32068997
  • Cas Number : 1590372-46-5
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-oxospiro[2.5]octane-1-carboxylate
Inchi Code 1S/C10H14O3/c1-13-9(12)8-6-10(8)4-2-7(11)3-5-10/h8H,2-6H2,1H3
Inchi Key JKCQCZMYZXVEDY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437603
Availability details for
Quantity
  • Availability
  • Add to Cart