ENAH5802EB81 Enamine

2-(oxan-2-yl)cyclohexan-1-one

  • MDL number MFCD31415388
  • Cas Number : 37749-86-3
  • Molecular Weight : 182.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426559
Availability details for
Quantity
  • Availability
  • Add to Cart