ENAH5802EB87 Enamine

ethyl 3-oxocycloheptane-1-carboxylate

  • MDL number MFCD16036069
  • Cas Number : 50559-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-217984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217984
Availability details for
Quantity
  • Availability
  • Add to Cart