ENAH5802EB88 Enamine

ethyl 2-oxo-2-(1,2-thiazol-5-yl)acetate

  • MDL number MFCD25962305
  • Cas Number : 32540-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1167108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1167108
Availability details for
Quantity
  • Availability
  • Add to Cart