ENAH5802EB8A Enamine

methyl 2-methoxy-2-(2-oxocyclopentyl)acetate

  • MDL number MFCD32066010
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methoxy-2-(2-oxocyclopentyl)acetate
Inchi Code 1S/C9H14O4/c1-12-8(9(11)13-2)6-4-3-5-7(6)10/h6,8H,3-5H2,1-2H3
Inchi Key WBQORVIVQHYSJE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7356764

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7356764
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart