ENAH5802EB8F Enamine

4,4,7-trimethyl-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD11590851
  • Cas Number : 70358-65-5
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4,7-trimethyl-3,4-dihydro-1(2H)-naphthalenone
Inchi Code 1S/C13H16O/c1-9-4-5-11-10(8-9)12(14)6-7-13(11,2)3/h4-5,8H,6-7H2,1-3H3
Inchi Key BANHFMVTHCHWSC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7356298

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7356298
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart