ENAH5802EB93 Enamine

methyl 3-oxo-2,3-dihydro-1-benzofuran-6-carboxylate

  • MDL number MFCD20462560
  • Cas Number : 1337858-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355428
Availability details for
Quantity
  • Availability
  • Add to Cart