ENAH5802EB94 Enamine

methyl 2-(4-acetylphenyl)acetate

  • MDL number MFCD09266207
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (4-acetylphenyl)acetate
Inchi Code 1S/C11H12O3/c1-8(12)10-5-3-9(4-6-10)7-11(13)14-2/h3-6H,7H2,1-2H3
Inchi Key PYSYDGVIWZCWSC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4271457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4271457
Availability details for
Quantity
  • Availability
  • Add to Cart