ENAH5802EB96 Enamine

6-(4-oxopiperidin-1-yl)-3,4-dihydropyrimidin-4-one hydrochloride

  • MDL number MFCD31924499
  • Cas Number : 2344680-04-0
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(6-hydroxypyrimidin-4-yl)piperidin-4-one hydrochloride
Inchi Code 1S/C9H11N3O2.ClH/c13-7-1-3-12(4-2-7)8-5-9(14)11-6-10-8;/h5-6H,1-4H2,(H,10,11,14);1H
Inchi Key NGSWZPMMGPRONT-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7424857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424857
Availability details for
Quantity
  • Availability
  • Add to Cart