ENAH5802EB9E Enamine

1,2-bis(pyridin-2-yl)ethan-1-one

  • MDL number MFCD21265114
  • Cas Number : 59576-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-2888679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2888679
Availability details for
Quantity
  • Availability
  • Add to Cart