ENAH5802EBA0 Enamine

2-(2,3-dihydro-1H-1,3-benzodiazol-2-ylidene)-3-oxobutanenitrile

  • MDL number MFCD00966509
  • Cas Number : 301344-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41511
Availability details for
Quantity
  • Availability
  • Add to Cart