ENAH5802EBA2 Enamine

1-(1-benzyl-1H-pyrrol-2-yl)ethan-1-one

  • MDL number MFCD00661628
  • Cas Number : 100713-02-8
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1-benzyl-1H-pyrrol-2-yl)ethan-1-one
Inchi Code 1S/C13H13NO/c1-11(15)13-8-5-9-14(13)10-12-6-3-2-4-7-12/h2-9H,10H2,1H3
Inchi Key ZXKCNDMXROKKRO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6999764

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6999764
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart