ENAH5802EBA4 Enamine

1-[3-(methylsulfanyl)propanoyl]piperidin-4-one

  • MDL number MFCD19595675
  • Cas Number : 1342451-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5292551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5292551
Availability details for
Quantity
  • Availability
  • Add to Cart