ENAH5802EBA5 Enamine

1-(2,2-dimethylthiomorpholin-4-yl)butan-2-one

  • MDL number MFCD32068913
  • Cas Number : 1595574-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5324008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5324008
Availability details for
Quantity
  • Availability
  • Add to Cart