ENAH5802EBA6 Enamine

4-(2,2-dimethylthiomorpholin-4-yl)butan-2-one

  • MDL number MFCD29932806
  • Cas Number : 1600592-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5033990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5033990
Availability details for
Quantity
  • Availability
  • Add to Cart