ENAH5802EBB1 Enamine

4-(trifluoroacetyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD24107290
  • Cas Number : 1514015-89-4
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2,2,2-trifluoroacetyl)-1H-pyrrole-2-carboxylic acid
Inchi Code 1S/C7H4F3NO3/c8-7(9,10)5(12)3-1-4(6(13)14)11-2-3/h1-2,11H,(H,13,14)
Inchi Key CNYWIHOEMYEBDC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5175128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5175128
Availability details for
Quantity
  • Availability
  • Add to Cart