ENAH5802EBB2 Enamine

5-methoxy-5-(trifluoromethyl)bicyclo[2.2.1]heptan-2-one

  • MDL number MFCD32066669
  • Cas Number : 2361644-59-7
  • Molecular Weight : 208.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S,4S,5R)-5-methoxy-5-(trifluoromethyl)bicyclo[2.2.1]heptan-2-one
Inchi Code 1S/C9H11F3O2/c1-14-8(9(10,11)12)4-5-2-6(8)3-7(5)13/h5-6H,2-4H2,1H3
Inchi Key CYFGELYIECADFM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7429915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429915
Availability details for
Quantity
  • Availability
  • Add to Cart