ENAH5802EBB4 Enamine

5,5-dimethyl-4-oxo-4,5,6,7-tetrahydro-1-benzofuran-2-carboxylic acid

  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,5-dimethyl-4-oxo-4,5,6,7-tetrahydrobenzofuran-2-carboxylic acid
Inchi Code 1S/C11H12O4/c1-11(2)4-3-7-6(9(11)12)5-8(15-7)10(13)14/h5H,3-4H2,1-2H3,(H,13,14)
Inchi Key GCPBTVFLLWEDFE-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form powder
Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430949
Availability details for
Quantity
  • Availability
  • Add to Cart