ENAH5802EBB5 Enamine

2-(4-fluorophenoxy)cyclohexan-1-one

  • MDL number MFCD22371312
  • Cas Number : 59798-93-5
  • Molecular Weight : 208.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431944
Availability details for
Quantity
  • Availability
  • Add to Cart