ENAH5802EBB9 Enamine

2-methyl-4-(3-methylbenzoyl)pyridine hydrochloride

  • MDL number MFCD32069399
  • Cas Number : 2361678-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435493
Availability details for
Quantity
  • Availability
  • Add to Cart