ENAH5802EBBC Enamine

2-methyl-5-(3-methylbenzoyl)pyridine hydrochloride

  • MDL number MFCD32068792
  • Cas Number : 2375269-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435925
Availability details for
Quantity
  • Availability
  • Add to Cart