ENAH5802EBC4 Enamine

4-(4-chlorophenyl)-2-oxobutanoic acid

  • MDL number MFCD19262228
  • Cas Number : 76400-21-0
  • Molecular Weight : 212.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-chlorophenyl)-2-oxobutanoic acid
Inchi Code 1S/C10H9ClO3/c11-8-4-1-7(2-5-8)3-6-9(12)10(13)14/h1-2,4-5H,3,6H2,(H,13,14)
Inchi Key HNZMKKXVFZVKII-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4260167

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-4260167
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart