ENAH5802EBC7 Enamine

2-(4-fluorobenzoyl)aniline

  • MDL number MFCD06658166
  • Cas Number : 3800-06-4
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6479199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6479199
Availability details for
Quantity
  • Availability
  • Add to Cart