ENAH5802EBD1 Enamine

methyl 4-(3-chlorophenyl)-4-oxobutanoate

  • MDL number MFCD11617551
  • Cas Number : 1249761-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-307734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-307734
Availability details for
Quantity
  • Availability
  • Add to Cart