ENAH5802EBD3 Enamine

1-(2-fluorophenyl)-3-phenylpropan-2-one

  • MDL number MFCD11938431
  • Cas Number : 1494-55-9
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 36-38
Iupac Name 1-(2-fluorophenyl)-3-phenylacetone
Inchi Code 1S/C15H13FO/c16-15-9-5-4-8-13(15)11-14(17)10-12-6-2-1-3-7-12/h1-9H,10-11H2
Inchi Key VVOSMBWUOVNUMM-UHFFFAOYSA-N

Description

Melting Point 36-38
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7354339

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7354339
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart