ENAH5802EBD4 Enamine

1-{1H,2H,3H,4H,9H-pyrido[3,4-b]indol-1-yl}propan-2-one hydrochloride

  • MDL number MFCD30526861
  • Cas Number : 157103-26-9
  • Molecular Weight : 264.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indol-1-yl)propan-2-one hydrochloride
Inchi Code 1S/C14H16N2O.ClH/c1-9(17)8-13-14-11(6-7-15-13)10-4-2-3-5-12(10)16-14;/h2-5,13,15-16H,6-8H2,1H3;1H
Inchi Key LABKMMWJWGDIIW-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7439263

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7439263
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart