ENAH5802EBD5 Enamine

7-methoxy-1,2,3,9,10,10a-hexahydrophenanthren-3-one

  • MDL number MFCD32198338
  • Cas Number : 65817-04-1
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-methoxy-1,9,10,10a-tetrahydrophenanthren-3(2H)-one

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6767141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6767141
Availability details for
Quantity
  • Availability
  • Add to Cart