ENAH5802EBD6 Enamine

ethyl N-[3-(2-oxopropyl)-1,2,4-thiadiazol-5-yl]carbamate

  • MDL number MFCD29993311
  • Cas Number : 64572-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434006
Availability details for
Quantity
  • Availability
  • Add to Cart