ENAH5802EBDA Enamine

1-(2,3,8,8-tetramethyl-1,2,3,4,5,6,7,8-octahydronaphthalen-2-yl)ethan-1-one

  • MDL number MFCD00217013
  • Cas Number : 54464-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430612
Availability details for
Quantity
  • Availability
  • Add to Cart