ENAH5802EBDC Enamine

tert-butyl 7-oxo-2-azaspiro[4.4]nonane-2-carboxylate

  • MDL number MFCD23106459
  • Cas Number : 1319716-42-1
  • Molecular Weight : 239.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1668513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1668513
Availability details for
Quantity
  • Availability
  • Add to Cart