ENAH5802EBDE Enamine

tert-butyl N-({3-oxospiro[3.3]heptan-1-yl}methyl)carbamate

  • MDL number MFCD32066714
  • Cas Number : 2361644-49-5
  • Molecular Weight : 239.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl ((3-oxospiro[3.3]heptan-1-yl)methyl)carbamate
Inchi Code 1S/C13H21NO3/c1-12(2,3)17-11(16)14-8-9-7-10(15)13(9)5-4-6-13/h9H,4-8H2,1-3H3,(H,14,16)
Inchi Key GYELLXXRNBKJBX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433319
Availability details for
Quantity
  • Availability
  • Add to Cart