ENAH5802EBDF Enamine

1-[(4-chlorophenyl)methoxy]piperidin-4-one

  • MDL number MFCD32069071
  • Cas Number : 2375267-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437782
Availability details for
Quantity
  • Availability
  • Add to Cart