ENAH5802EBE3 Enamine

2-[2-(2,2-dimethylthiomorpholin-4-yl)ethyl]cyclopentan-1-one

  • Cas Number : 1594792-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7442532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442532
Availability details for
Quantity
  • Availability
  • Add to Cart