ENAH5802EBE8 Enamine

4-(3,4-dichlorophenyl)-2-oxobutanoic acid

  • MDL number MFCD19262231
  • Cas Number : 85918-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7432570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432570
Availability details for
Quantity
  • Availability
  • Add to Cart