ENAH5802EBEB Enamine

tert-butyl 2-(difluoromethyl)-4-oxopiperidine-1-carboxylate

  • MDL number MFCD28343805
  • Cas Number : 362705-39-3
  • Molecular Weight : 249.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-(difluoromethyl)-4-oxopiperidine-1-carboxylate
Inchi Code 1S/C11H17F2NO3/c1-11(2,3)17-10(16)14-5-4-7(15)6-8(14)9(12)13/h8-9H,4-6H2,1-3H3
Inchi Key RIJDMPSJVNBKJV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7054368

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7054368
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart