ENAH5802EBEC Enamine

tert-butyl N-{2-oxospiro[3.5]nonan-7-yl}carbamate

  • MDL number MFCD24857292
  • Cas Number : 1440962-35-5
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (2-oxospiro[3.5]nonan-7-yl)carbamate
Inchi Code 1S/C14H23NO3/c1-13(2,3)18-12(17)15-10-4-6-14(7-5-10)8-11(16)9-14/h10H,4-9H2,1-3H3,(H,15,17)
Inchi Key SPZQOGMHBFODOY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7439170

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7439170
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart