ENAH5802EBED Enamine

2-(4-butylbenzoyl)aniline

  • Cas Number : 1225788-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25746
Availability details for
Quantity
  • Availability
  • Add to Cart